3-Fluoro-5-methylbenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Fluoro-5-methylphenyl)methanol

PC5916-1G PC5916-5G PC5916-10G
APOSPC5916-1GEA 46.4 EUR
APOSPC5916-1G APOSPC5916-5G APOSPC5916-10G
3-Fluoro-5-methylbenzyl alcohol
3-Fluoro-5-methylbenzyl alcohol
Formel: C₈H₉FO
Molecular Weight: 140.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094314
CAS-Nummer: 518070-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR