3-Fluoro-5-methylbenzamide 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC5922-5G
PC5922-1G
APOSPC5922-5GEA
335
EUR
APOSPC5922-5G
APOSPC5922-1G
3-Fluoro-5-methylbenzamide 97%
3-Fluoro-5-methylbenzamide
Formel:
C₈H₈FNO Molecular Weight: 153,154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094319 CAS-Nummer: 518070-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...