4-(Bromomethyl)-2-fluorobenzonitrile 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Cyano-3-fluorobenzyl bromide, α-Bromo-2-fluoro-p-tolunitrile

PC5928-1G PC5928-25G PC5928-5G PC5928-10G
APOSPC5928-1GEA 39.3 EUR
APOSPC5928-1G APOSPC5928-25G APOSPC5928-5G APOSPC5928-10G
4-(Bromomethyl)-2-fluorobenzonitrile 98%
4-(Bromomethyl)-2-fluorobenzonitrile
Formel: C₈H₅BrFN
Molecular Weight: 214,034 g/mol
Schmelzpunkt: 44…46 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03094323
CAS-Nummer: 222978-03-2
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR