Perfluorocyclohexanecarbonyl fluoride 65%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Undecafluorocyclohexanecarbonyl fluoride, (Fluorocarbonyl)perfluorocyclohexane

PC5932E-10G PC5932E-50G
APOSPC5932E-10GEA 21.9 EUR
APOSPC5932E-10G APOSPC5932E-50G
Perfluorocyclohexanecarbonyl fluoride 65%
Perfluorocyclohexanecarbonyl fluoride
Formel: C₇F₁₂O
Siedepunkt: 70 °C
Dichte: 1,6 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00236697
CAS-Nummer: 6588-63-2
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR