2-Fluoro-4-methylbenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-(hydroxymethyl)toluene, (2-Fluoro-4-methylphenyl)methanol

PC5935-1G PC5935-5G PC5935-25G
APOSPC5935-1GEA 36.4 EUR
APOSPC5935-1G APOSPC5935-5G APOSPC5935-25G
2-Fluoro-4-methylbenzyl alcohol
2-Fluoro-4-methylbenzyl alcohol
Formel: C₈H₉FO
Molecular Weight: 140,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094326
CAS-Nummer: 252004-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR