1H,6H-Dodecafluorohexane 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,1,2,2,3,3,4,4,5,5,6,6-Dodecafluorohexane
PC5943-25G
PC5943-5G
PC5943-1G
APOSPC5943-25GEA
430
EUR
APOSPC5943-25G
APOSPC5943-5G
APOSPC5943-1G
1H,6H-Dodecafluorohexane 97%
1H,6H-Dodecafluorohexane
Formel:
C₆H₂F₁₂ Molecular Weight: 302,061 g/mol Siedepunkt: 92,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155777 CAS-Nummer: 336-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...