1H,2H-Perfluoro-1-iodooct-1-ene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H,2H,1-Iodoperfluorooct-1-ene

PC5984-1G PC5984-250MG PC5984-5G
APOSPC5984-1GEA 236 EUR
APOSPC5984-1G APOSPC5984-250MG APOSPC5984-5G
1H,2H-Perfluoro-1-iodooct-1-ene
1H,2H-Perfluoro-1-iodooct-1-ene
Formel: C₈H₂F₁₃I
Molecular Weight: 471,985 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094337
CAS-Nummer: 150223-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR