4-Amino-5-chloro-2,6-difluoropyrimidine, tech
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2,6-difluoropyrimidin-4-amine
PC5989-1G
PC5989-250MG
APOSPC5989-1GEA
185
EUR
APOSPC5989-1G
APOSPC5989-250MG
4-Amino-5-chloro-2,6-difluoropyrimidine, tech
4-Amino-5-chloro-2,6-difluoropyrimidine
Contains up to 15% 2-Amino-5-chloro-4,6-difluoropyrimidine
Formel:
C₄H₂ClF₂N₃ Molecular Weight: 165,529 g/mol Schmelzpunkt: 158…164 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13563124 CAS-Nummer: 27078-72-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...