Ethyl 3-chloro-4-fluorobenzoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-4-fluorobenzoic acid ethyl ester
PC6017-1G
PC6017-5G
PC6017-25G
APOSPC6017-1GEA
14.7
EUR
APOSPC6017-1G
APOSPC6017-5G
APOSPC6017-25G
Ethyl 3-chloro-4-fluorobenzoate
Ethyl 3-chloro-4-fluorobenzoate
Formel:
C₉H₈ClFO₂ Molecular Weight: 202,61 g/mol Schmelzpunkt: 39…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204341 CAS-Nummer: 137521-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...