4-Bromo-3-(trifluoromethyl)phenylacetonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-(cyanomethyl)benzotrifluoride, 4-Bromo-3-(trifluoromethyl)benzyl cyanide

PC6026-1G PC6026-5G
APOSPC6026-1GEA 132 EUR
APOSPC6026-1G APOSPC6026-5G
4-Bromo-3-(trifluoromethyl)phenylacetonitrile
4-Bromo-3-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₅BrF₃N
Molecular Weight: 264,042 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026483
CAS-Nummer: 1159512-69-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR