3,5-Difluoro-4-methoxybenzyl alcohol 97%

Lieferant: Apollo Scientific

Synonyme: 2,6-Difluoro-4-(hydroxymethyl)anisole, 3,5-Difluoro-4-methoxybenzyl Alcohol

PC6031-10G PC6031-5G
APOSPC6031-10GEA 237 EUR
APOSPC6031-10G APOSPC6031-5G
3,5-Difluoro-4-methoxybenzyl alcohol 97%
3,5-Difluoro-4-methoxybenzyl alcohol
Formel: C₈H₈F₂O₂
Molecular Weight: 174.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115914
CAS-Nummer: 706786-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR