2H,3H,3H-Perfluorobut-1-ene

Lieferant: Apollo Scientific

PC6047-25G PC6047-5G
APOSPC6047-25GEA 1360 EUR
APOSPC6047-25G APOSPC6047-5G
2H,3H,3H-Perfluorobut-1-ene
2H,3H,3H-Perfluorobut-1-ene
Formel: C₄H₃F₅
Lagertemperatur: Raumtemperatur
MDL: MFCD12922835
CAS-Nummer: 721946-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR