2-[(2-Fluorophenyl)amino]aniline 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N1-(2-Fluorophenyl)benzene-1,2-diamine, 2-Amino-2'-fluorodiphenylamine
PC6058-1G
PC6058-25G
PC6058-5G
APOSPC6058-1GEA
23.2
EUR
APOSPC6058-1G
APOSPC6058-25G
APOSPC6058-5G
2-[(2-Fluorophenyl)amino]aniline 98%
2-[(2-Fluorophenyl)amino]aniline
Formel:
C₁₂H₁₁FN₂ Molecular Weight: 202,228 g/mol Schmelzpunkt: 73…75 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10567164 CAS-Nummer: 28898-03-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...