3-Fluoro-4-(methylsulphonyl)benzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Cyano-2-fluorophenyl methyl sulphone
PC6059-1G
PC6059-5G
APOSPC6059-1GEA
132
EUR
APOSPC6059-1G
APOSPC6059-5G
3-Fluoro-4-(methylsulphonyl)benzonitrile
3-Fluoro-4-(methylsulphonyl)benzonitrile
Formel:
C₈H₆FNO₂S Molecular Weight: 199.202 g/mol Schmelzpunkt: 135…137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12922838 CAS-Nummer: 185946-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...