3-Perfluorohexyl-2-iodopropan-1-ol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC6088D-1G
PC6088D-5G
APOSPC6088D-1GEA
116
EUR
APOSPC6088D-1G
APOSPC6088D-5G
3-Perfluorohexyl-2-iodopropan-1-ol 97%
3-Perfluorohexyl-2-iodopropan-1-ol
Formel:
C₉H₆F₁₃IO Molecular Weight: 504,027 g/mol Siedepunkt: 90 °C (1,5 mm Hg) Schmelzpunkt: 49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236070 CAS-Nummer: 38550-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...