5-(Bromomethyl)-2-fluorobenzonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Cyano-4-fluorobenzyl bromide, α-Bromo-6-fluoro-m-tolunitrile
PC6134-5G
PC6134-1G
PC6134-25G
PC6134-100G
APOSPC6134-5GEA
79.9
EUR
APOSPC6134-5G
APOSPC6134-1G
APOSPC6134-25G
APOSPC6134-100G
5-(Bromomethyl)-2-fluorobenzonitrile
5-(Bromomethyl)-2-fluorobenzonitrile
Formel:
C₈H₅BrFN Molecular Weight: 214.034 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094341 CAS-Nummer: 180302-35-6 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...