2-Perfluoroethyl-1H-benzimidazole 98%

Lieferant: Apollo Scientific

Synonyme: 2-(Pentafluoroethyl)-1H-benzimidazole

PC6178-1G PC6178-5G
APOSPC6178-1GEA 198 EUR
APOSPC6178-1G APOSPC6178-5G
2-Perfluoroethyl-1H-benzimidazole 98%
2-Perfluoroethyl-1H-benzimidazole
Formel: C₉H₅F₅N₂
Molecular Weight: 236,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779575
CAS-Nummer: 383-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR