4'-(Trifluoroacetyl)acetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Acetylphenyl)-2,2,2-trifluoroethan-1-one, 1-(4-Acetylphenyl)-2,2,2-trifluoroethanone
PC6195-1G
PC6195-250MG
PC6195-5G
APOSPC6195-1GEA
192
EUR
APOSPC6195-1G
APOSPC6195-250MG
APOSPC6195-5G
4'-(Trifluoroacetyl)acetophenone
4'-(Trifluoroacetyl)acetophenone
Formel:
C₁₀H₇F₃O₂ Molecular Weight: 216,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12547944 CAS-Nummer: 86988-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...