Tris(perfluorohex-1-yl)amine, tech
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Tris(tridecafluorohex-1-yl)amine
PC6220G-1G
PC6220G-5G
APOSPC6220G-1GEA
80.3
EUR
APOSPC6220G-1G
APOSPC6220G-5G
Tris(perfluorohex-1-yl)amine, tech
Tris(perfluorohex-1-yl)amine
Formel:
C₁₈F₃₉N Molecular Weight: 971.137 g/mol Siedepunkt: 250…260 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00042590 CAS-Nummer: 432-08-6 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...