4-Bromo-2-fluorophenyl methyl sulphone 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Bromo-2-fluoro-1-(methylsulphonyl)benzene

PC6251-1G PC6251-5G
APOSPC6251-1GEA 87 EUR
APOSPC6251-1G APOSPC6251-5G
4-Bromo-2-fluorophenyl methyl sulphone 98%
4-Bromo-2-fluorophenyl methyl sulphone
Formel: C₇H₆BrFO₂S
Molecular Weight: 253.089 g/mol
Schmelzpunkt: 91…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11855948
CAS-Nummer: 648904-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR