3-Fluoro-4-(methylsulphonyl)benzaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-formylphenyl methyl sulphone
PC6278-1G
PC6278-250MG
PC6278-5G
APOSPC6278-1GEA
244
EUR
APOSPC6278-1G
APOSPC6278-250MG
APOSPC6278-5G
3-Fluoro-4-(methylsulphonyl)benzaldehyde 97%
3-Fluoro-4-(methylsulphonyl)benzaldehyde
Formel:
C₈H₇FO₃S Schmelzpunkt: 122…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846869 CAS-Nummer: 254878-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...