2-Fluoro-3-(trifluoromethoxy)phenylacetonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Cyanomethyl)-α,α,α,2-tetrafluoroanisole, 2-Fluoro-3-(trifluoromethoxy)benzyl cyanide

PC6351-1G PC6351-5G PC6351-10G
APOSPC6351-1GEA 192 EUR
APOSPC6351-1G APOSPC6351-5G APOSPC6351-10G
2-Fluoro-3-(trifluoromethoxy)phenylacetonitrile
2-Fluoro-3-(trifluoromethoxy)phenylacetonitrile
Formel: C₉H₅F₄NO
Molecular Weight: 219,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026475
CAS-Nummer: 1159512-60-3
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR