2-Fluoro-5-iodobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Fluoro-5-iodophenyl)methanol

PC6376-1G PC6376-250MG PC6376-5G
APOSPC6376-1GEA 48 EUR
APOSPC6376-1G APOSPC6376-250MG APOSPC6376-5G
2-Fluoro-5-iodobenzyl alcohol
2-Fluoro-5-iodobenzyl alcohol
Formel: C₇H₆FIO
Molecular Weight: 252.025 g/mol
Schmelzpunkt: 42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06202824
CAS-Nummer: 438050-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR