2'-Fluoro-5'-iodoacetophenone 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-5-iodophenyl)ethan-1-one, 1-(2-Fluoro-5-iodophenyl)ethanone

PC6378-1G PC6378-5G
APOSPC6378-1GEA 65.4 EUR
APOSPC6378-1G APOSPC6378-5G
2'-Fluoro-5'-iodoacetophenone 99%
2'-Fluoro-5'-iodoacetophenone
Formel: C₈H₆FIO
Molecular Weight: 264,036 g/mol
Schmelzpunkt: 46…46,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12026480
CAS-Nummer: 1159512-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR