4-Methoxy-2-(trifluoromethoxy)benzyl alcohol 95%

Lieferant: Apollo Scientific

Synonyme: [4-Methoxy-2-(trifluoromethoxy)phenyl]methanol

PC6388-1G PC6388-250MG
APOSPC6388-1GEA 144 EUR
APOSPC6388-1G APOSPC6388-250MG
4-Methoxy-2-(trifluoromethoxy)benzyl alcohol 95%
4-Methoxy-2-(trifluoromethoxy)benzyl alcohol
Formel: C₉H₉F₃O₃
Molecular Weight: 222,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660288
CAS-Nummer: 886502-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR