3,5-Difluoro-4-methoxybenzoyl chloride 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(Chloroformyl)-2,6-difluoroanisole, 4-(Chlorocarbonyl)-2,6-difluoroanisole
PC6393-1G
PC6393-5G
APOSPC6393-1GEA
56.7
EUR
APOSPC6393-1G
APOSPC6393-5G
3,5-Difluoro-4-methoxybenzoyl chloride 98%
3,5-Difluoro-4-methoxybenzoyl chloride
Formel:
C₈H₅ClF₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115913 CAS-Nummer: 501701-43-5 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...