4-Chloro-1,1,1,3,3-pentafluorobutane 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chloro-1H,1H,3H,3H-perfluorobutane
PC6399-25G
PC6399-5G
APOSPC6399-25GEA
575
EUR
APOSPC6399-25G
APOSPC6399-5G
4-Chloro-1,1,1,3,3-pentafluorobutane 97%
4-Chloro-1,1,1,3,3-pentafluorobutane
Formel:
C₄H₄ClF₅ Molecular Weight: 182,52 g/mol Siedepunkt: 78 °C (760 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784191 CAS-Nummer: 70566-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...