3-Hydroxy-4,4,4-trifluorobutanoic acid 97+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Hydroxy-4,4,4-trifluorobutyric acid, 3-Hydroxy-3-(trifluoromethyl)propanoic acid
PC6418-1G
PC6418-5G
APOSPC6418-1GEA
84.6
EUR
APOSPC6418-1G
APOSPC6418-5G
3-Hydroxy-4,4,4-trifluorobutanoic acid 97+%
3-Hydroxy-4,4,4-trifluorobutanoic acid
Formel:
C₄H₅F₃O₃ Molecular Weight: 158.076 g/mol Schmelzpunkt: 78…79 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00088012 CAS-Nummer: 86884-21-1 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...