Sodium perfluoropropanoate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Pentafluoropropionic acid sodium salt, Sodium pentafluoropropionate
PC6540-100G
PC6540-25G
APOSPC6540-100GEA
364
EUR
APOSPC6540-100G
APOSPC6540-25G
Sodium perfluoropropanoate 98%
Sodium perfluoropropanoate
Formel:
C₃F₅O₂·Na Molecular Weight: 186,013 g/mol Schmelzpunkt: 226…229 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066167 CAS-Nummer: 378-77-8 EINECS: 206-830-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...