5-Chloro-2-nitro-4-(trifluoromethyl)phenylhydrazine

Lieferant: Apollo Scientific

PC6607-1G PC6607-250MG
APOSPC6607-1GEA 244 EUR
APOSPC6607-1G APOSPC6607-250MG
5-Chloro-2-nitro-4-(trifluoromethyl)phenylhydrazine
5-Chloro-2-nitro-4-(trifluoromethyl)phenylhydrazine
Formel: C₇H₅ClF₃N₃O₂
Molecular Weight: 255,582 g/mol
Schmelzpunkt: 139…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00119559
CAS-Nummer: 306935-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR