1-(4-Fluorophenyl)propan-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Fluorophenyl)-1-hydroxypropane, α-Ethyl-4-fluorobenzyl alcohol
PC6642-1G
PC6642-5G
APOSPC6642-1GEA
126
EUR
APOSPC6642-1G
APOSPC6642-5G
1-(4-Fluorophenyl)propan-1-ol
1-(4-Fluorophenyl)propan-1-ol
Formel:
C₉H₁₁FO Molecular Weight: 154,181 g/mol Siedepunkt: 116 °C (17 mm Hg) Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021912 CAS-Nummer: 701-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...