6-[4-(Trifluoromethyl)phenyl]-1,3,5-triazine-2,4-diamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,4-Diamino-6-[(4-(trifluoromethyl)phenyl)]-1,3,5-triazine, 4-(4,6-Diamino-1,3,5-triazin-2-yl)benzotrifluoride

PC6644-25G PC6644-5G PC6644-100G
APOSPC6644-25GEA 261 EUR
APOSPC6644-25G APOSPC6644-5G APOSPC6644-100G
6-[4-(Trifluoromethyl)phenyl]-1,3,5-triazine-2,4-diamine
6-[4-(Trifluoromethyl)phenyl]-1,3,5-triazine-2,4-diamine
Formel: C₁₀H₈F₃N₅
Molecular Weight: 255,199 g/mol
Schmelzpunkt: 239…241 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00268940
CAS-Nummer: 186834-97-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR