3-Amino-1,1,1-trifluoropropan-2-ol hydrochloride 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-3,3,3-trifluoropropylamine hydrochloride

PC6662-1G PC6662-250MG
APOSPC6662-1GEA 143 EUR
APOSPC6662-1G APOSPC6662-250MG
3-Amino-1,1,1-trifluoropropan-2-ol hydrochloride 97+%
3-Amino-1,1,1-trifluoropropan-2-ol hydrochloride
Formel: C₃H₆F₃NO·ClH
Molecular Weight: 165,542 g/mol
Schmelzpunkt: 154…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784296
CAS-Nummer: 3832-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR