(Z)-1-Bromo-2-fluoroethylene 97%

Lieferant: Apollo Scientific
Achtung

PC6672-25G PC6672-5G
APOSPC6672-25GEA 894 EUR
APOSPC6672-25G APOSPC6672-5G
(Z)-1-Bromo-2-fluoroethylene 97%
(Z)-1-Bromo-2-fluoroethylene
Formel: C₂H₂BrF
Molecular Weight: 124,94 g/mol
Siedepunkt: 36 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20543004
CAS-Nummer: 2366-31-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR