2-Acetyl-3-amino-5-(4-fluorophenyl)thiophene 97%

Lieferant: Apollo Scientific

PC6701-1G PC6701-2.5G
APOSPC6701-1GEA 72.7 EUR
APOSPC6701-1G APOSPC6701-2.5G
2-Acetyl-3-amino-5-(4-fluorophenyl)thiophene 97%
2-Acetyl-3-amino-5-(4-fluorophenyl)thiophene
Formel: C₁₂H₁₀FNOS
Molecular Weight: 235,277 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179905
CAS-Nummer: 306935-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR