[(S,S)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)]difluorotitanium(IV)

Lieferant: Apollo Scientific
Gefahr

PC6741-100MG PC6741-25MG
APOSPC6741-100MGEA 198 EUR
APOSPC6741-100MG APOSPC6741-25MG
[(S,S)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)]difluorotitanium(IV)
[(S,S)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)]difluorotitanium(IV)

Precatalyst for the highly enantioselective imine hydrosilylation

Formel: C₂₀H₂₄F₂Ti
Molecular Weight: 350,268 g/mol
Schmelzpunkt: 230 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01863622
CAS-Nummer: 178177-04-3
UN: 1759
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR