4-Amino-5-(4-fluorophenyl)-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-2,4-dihydro-5-(4-fluorophenyl)-3H-1,2,4-triazole-3-thione, 4-Amino-3-(4-fluorophenyl)-5-mercapto-4H-1,2,4-triazole

PC6800-1G PC6800-100MG
APOSPC6800-1GEA 163 EUR
APOSPC6800-1G APOSPC6800-100MG
4-Amino-5-(4-fluorophenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(4-fluorophenyl)-4H-1,2,4-triazole-3-thiol
Formel: C₈H₇FN₄S
Molecular Weight: 210,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269268
CAS-Nummer: 61019-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR