1H-Perfluoro(cyclopent-1-ene) 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3,3,4,4,5,5-Heptafluorocyclopent-1-ene

PC6813-1G PC6813-250MG
APOSPC6813-1GEA 335 EUR
APOSPC6813-1G APOSPC6813-250MG
1H-Perfluoro(cyclopent-1-ene) 97%
1H-Perfluoro(cyclopent-1-ene)
Formel: C₅HF₇
Molecular Weight: 194.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784216
CAS-Nummer: 1892-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR