Methyl 3,5,6-trichloroperfluorohexanoate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3,5,6-trichlorooctafluorohexanoate
PC6829-1G
PC6829-250MG
PC6829-5G
APOSPC6829-1GEA
219
EUR
APOSPC6829-1G
APOSPC6829-250MG
APOSPC6829-5G
Methyl 3,5,6-trichloroperfluorohexanoate 95%
Methyl 3,5,6-trichloroperfluorohexanoate
Formel:
C₇H₃Cl₃F₈O₂ Molecular Weight: 377,444 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10567158 CAS-Nummer: 812-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...