Methyl 4-bromo-2,2,3,3,4,4-hexafluorobutanoate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Methyl perfluoro-4-bromobutanoate, Methyl 4-bromo-2,2,3,3,4,4-hexafluorobutyrate
PC6835-1G
PC6835-250MG
APOSPC6835-1GEA
306
EUR
APOSPC6835-1G
APOSPC6835-250MG
Methyl 4-bromo-2,2,3,3,4,4-hexafluorobutanoate
Methyl 4-bromo-2,2,3,3,4,4-hexafluorobutanoate
Formel:
C₅H₃BrF₆O₂ Molecular Weight: 288.971 g/mol Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09763602 CAS-Nummer: 128160-31-6 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...