2,3,3,3-Tetrafluoropropionamide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC6866-10G
PC6866-25G
PC6866-5G
APOSPC6866-10GEA
379
EUR
APOSPC6866-10G
APOSPC6866-25G
APOSPC6866-5G
2,3,3,3-Tetrafluoropropionamide 97%
2,3,3,3-Tetrafluoropropionamide
Formel:
C₃H₃F₄NO Molecular Weight: 145,056 g/mol Schmelzpunkt: 52…54 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069119 CAS-Nummer: 1647-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...