1H,1H,2H,2H-Perfluorohexanesulphonic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,3,4,4,5,5,6,6,6-Nonafluorohexane-1-sulphonic acid

PC6901-1G PC6901-250MG
APOSPC6901-1GEA 720 EUR
APOSPC6901-1G APOSPC6901-250MG
1H,1H,2H,2H-Perfluorohexanesulphonic acid
1H,1H,2H,2H-Perfluorohexanesulphonic acid
Formel: C₆H₅F₉O₃S
Molecular Weight: 328,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14581696
CAS-Nummer: 757124-72-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR