3,6-Dibromo-2,4-difluorobenzaldehyde

Lieferant: Apollo Scientific
Achtung

PC6906-1G PC6906-5G
APOSPC6906-1GEA 141 EUR
APOSPC6906-1G APOSPC6906-5G
3,6-Dibromo-2,4-difluorobenzaldehyde
3,6-Dibromo-2,4-difluorobenzaldehyde
Formel: C₇H₂Br₂F₂O
Molecular Weight: 299,895 g/mol
Schmelzpunkt: 67…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11845983
CAS-Nummer: 1160573-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR