4-Fluoro-2-methyl-5-nitrobenzoic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Carboxy-2-fluoro-4-methylnitrobenzene, 2-Carboxy-5-fluoro-4-nitrotoluene

PC6925-1G PC6925-5G
APOSPC6925-1GEA 21.9 EUR
APOSPC6925-1G APOSPC6925-5G
4-Fluoro-2-methyl-5-nitrobenzoic acid 98%
4-Fluoro-2-methyl-5-nitrobenzoic acid
Formel: C₈H₆FNO₄
Molecular Weight: 199,136 g/mol
Schmelzpunkt: 211…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847453
CAS-Nummer: 64695-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR