6-Bromo-3-fluoropyridine-2-carboxaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-3-fluoropicolinaldehyde, 6-Bromo-3-fluoropyridine-2-carbaldehyde, 6-Bromo-3-fluoro-2-formylpyridine
PC6979-1G
PC6979-250MG
APOSPC6979-1GEA
84.1
EUR
APOSPC6979-1G
APOSPC6979-250MG
6-Bromo-3-fluoropyridine-2-carboxaldehyde 98%
6-Bromo-3-fluoropyridine-2-carboxaldehyde
Formel:
C₆H₃BrFNO Molecular Weight: 203,997 g/mol Schmelzpunkt: 114…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781224 CAS-Nummer: 885267-36-7 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...