2-Chloro-3-fluoroisonicotinaldehyde hydrate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-fluoropyridine-4-carboxaldehyde hydrate, 2-Chloro-3-fluoro-4-formylpyridine hydrate

PC7014-5G PC7014-100G PC7014-25G
APOSPC7014-5GEA 118 EUR
APOSPC7014-5G APOSPC7014-100G APOSPC7014-25G
2-Chloro-3-fluoroisonicotinaldehyde hydrate 98%
2-Chloro-3-fluoroisonicotinaldehyde hydrate
Formel: C₆H₃ClFNO·H₂O
Molecular Weight: 177,561 g/mol
Dichte: 0,936 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD06657826
CAS-Nummer: 1196156-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR