3-Fluoro-2-hydroxybenzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Borono-6-fluorophenol

PC7043-500MG PC7043-1G PC7043-5G
APOSPC7043-500MGEA 69.6 EUR
APOSPC7043-500MG APOSPC7043-1G APOSPC7043-5G
3-Fluoro-2-hydroxybenzeneboronic acid
3-Fluoro-2-hydroxybenzeneboronic acid
Formel: C₆H₆BFO₃
Molecular Weight: 155,919 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07781238
CAS-Nummer: 259209-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR