3-Fluoro-2-(morpholin-4-yl)pyridine

Lieferant: Apollo Scientific

PC7051-1G PC7051-5G PC7051-25G
APOSPC7051-1GEA 114 EUR
APOSPC7051-1G APOSPC7051-5G APOSPC7051-25G
3-Fluoro-2-(morpholin-4-yl)pyridine
3-Fluoro-2-(morpholin-4-yl)pyridine
Formel: C₉H₁₁FN₂O
Molecular Weight: 182,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781240
CAS-Nummer: 693235-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR