2-Cyano-4-(trifluoromethyl)benzeneboronic acid neopentyl glycol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-5-(trifluoromethyl)benzonitrile

PC7062-1G PC7062-250MG
APOSPC7062-1GEA 836 EUR
APOSPC7062-1G APOSPC7062-250MG
2-Cyano-4-(trifluoromethyl)benzeneboronic acid neopentyl glycol ester
2-Cyano-4-(trifluoromethyl)benzeneboronic acid neopentyl glycol ester
Formel: C₁₃H₁₃BF₃NO₂
Molecular Weight: 283,054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689554
CAS-Nummer: 883898-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR