1-(Trifluoromethyl)vinylboronic acid hexylene glycol ester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4,6-Trimethyl-2-(3,3,3-trifluoroprop-1-en-2-yl)-1,3,2-dioxaborinane
PC7067-1G
PC7067-5G
PC7067-250MG
APOSPC7067-1GEA
24.8
EUR
APOSPC7067-1G
APOSPC7067-5G
APOSPC7067-250MG
1-(Trifluoromethyl)vinylboronic acid hexylene glycol ester 98%
1-(Trifluoromethyl)vinylboronic acid hexylene glycol ester
Formel:
C₉H₁₄BF₃O₂ Molecular Weight: 222,012 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669623 CAS-Nummer: 1011460-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...